1-((3-Bromo-4-ethoxyphenyl)sulfonyl)-4-methylpiperazine 95%

Supplier: Ambeed
Warning

A188672-100MG A188672-1G A188672-250MG
77012-212EA 23 USD
77012-212 77012-214 77012-216
1-((3-Bromo-4-ethoxyphenyl)sulfonyl)-4-methylpiperazine 95%
1-((3-Bromo-4-ethoxyphenyl)sulfonyl)-4-methylpiperazine

Precaución: for Research Use Only

Formula: C₁₃H₁₉BrN₂O₃S
MDL Number: MFCD27923092
CAS Number: 1093065-11-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR