2-(4-Hydroxycyclohexyl)acetic acid 95%

Supplier: Ambeed
Warning

A188647-100MG A188647-1G A188647-250MG A188647-5G
77012-206EA 23.34 USD
77012-206 77012-208 77012-210 77683-483
2-(4-Hydroxycyclohexyl)acetic acid 95%
2-(4-Hydroxycyclohexyl)acetic acid

Precaución: For research use only.

Formula: C₈H₁₄O₃
MW: 158.20 g/mol
MDL Number: MFCD13659403
CAS Number: 99799-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR