(4-(Pyridin-3-yl)phenyl)boronic acid 97%

Supplier: Ambeed
Warning

A188598-250MG A188598-5G A188598-1G
76986-010EA 9 USD
76986-010 76986-012 77890-165
(4-(Pyridin-3-yl)phenyl)boronic acid 97%
(4-(Pyridin-3-yl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₀BNO₂
MW: 199.01 g/mol
MDL Number: MFCD18418033
CAS Number: 170230-28-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR