Methyl 2-chloro-3-methylbenzoate 98%

Supplier: Ambeed
Warning

A188437-100G A188437-1G A188437-25G A188437-5G
77029-732EA 331.01 USD
77029-732 77029-734 77029-736 77029-738
Methyl 2-chloro-3-methylbenzoate 98%
Methyl 2-chloro-3-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₉ClO₂
MW: 184.61 g/mol
MDL Number: MFCD11226202
CAS Number: 920759-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR