4-Bromo-5-chloro-1H-indazole 95%

Supplier: Ambeed
Warning

A188123-100MG A188123-1G A188123-250MG A188123-5G
77012-190EA 61.54 USD
77012-190 77012-192 77012-194 77012-196
4-Bromo-5-chloro-1H-indazole 95%
4-Bromo-5-chloro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄BrClN₂
MW: 231.4800 g/mol
MDL Number: MFCD12828060
CAS Number: 1056264-74-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR