4-Propylphenylacetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-(4-Propylphenyl)acetic acid

A188025-1G A188025-250MG A188025-5G
76998-898EA 12.74 USD
76998-898 76998-900 77671-668
4-Propylphenylacetic acid 97%
4-Propylphenylacetic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₂
MW: 178.228 g/mol
MDL Number: MFCD00600398
CAS Number: 26114-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR