Ethyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)nicotinate 96%

Supplier: Ambeed
Warning

A188012-1G A188012-25G A188012-5G
76985-996EA 36.08 USD
76985-996 76985-998 76986-000
Ethyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)nicotinate 96%
Ethyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)nicotinate

Precaución: for Research Use Only

Formula: C₁₄H₂₀BNO₄
MW: 277.12 g/mol
MDL Number: MFCD04117940
CAS Number: 916326-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR