1-(Pyrimidin-2-yl)-1H-pyrazole-4-carboxylic acid 95%

Supplier: Ambeed
Warning

A188011-100MG A188011-1G
76985-992EA 99.73 USD
76985-992 76985-994
1-(Pyrimidin-2-yl)-1H-pyrazole-4-carboxylic acid 95%
1-(Pyrimidin-2-yl)-1H-pyrazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₆N₄O₂
MW: 190.15 g/mol
MDL Number: MFCD12192706
CAS Number: 1014632-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR