6-Chloro-3-iodoimidazo[1,2-a]pyrazine 98%

Supplier: Ambeed
Warning

A187970-100MG A187970-250MG
77029-712EA 44.56 USD
77029-712 77029-714
6-Chloro-3-iodoimidazo[1,2-a]pyrazine 98%
6-Chloro-3-iodoimidazo[1,2-a]pyrazine

Precaución: for Research Use Only

Formula: C₆H₃ClIN₃
MW: 279.47 g/mol
MDL Number: MFCD18072709
CAS Number: 1245645-10-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR