Methyl-2-(2-amino-5-methyl-1,3-thiazol-4-yl)acetate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-(2-amino-5-methylthiazol-4-yl)acetate

A187967-1G A187967-250MG
77012-186EA 36.08 USD
77012-186 77012-188
Methyl-2-(2-amino-5-methyl-1,3-thiazol-4-yl)acetate 97%
Methyl-2-(2-amino-5-methyl-1,3-thiazol-4-yl)acetate

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O₂S
MW: 186.23 g/mol
MDL Number: MFCD00662517
CAS Number: 259654-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR