Ethyl 5-bromo-2-fluorobenzoate 97%

Supplier: Ambeed
Warning

A187884-1G A187884-25G A187884-5G
76985-986EA 32 USD
76985-986 76985-988 76985-990
Ethyl 5-bromo-2-fluorobenzoate 97%
Ethyl 5-bromo-2-fluorobenzoate

Precaución: for Research Use Only

Formula: C₉H₈BrFO₂
MDL Number: MFCD06204416
CAS Number: 612835-53-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR