4-Fluoro-3-[(trifluoromethyl)sulfonyl]benzenesulfonyl chloride 95%

Supplier: Ambeed
Danger

A187776-100MG A187776-1G A187776-250MG
77029-706EA 23 USD
77029-706 77029-708 77029-710
4-Fluoro-3-[(trifluoromethyl)sulfonyl]benzenesulfonyl chloride 95%
4-Fluoro-3-[(trifluoromethyl)sulfonyl]benzenesulfonyl chloride

Precaución: for Research Use Only

Formula: C₇H₃ClF₄O₄S₂
MDL Number: MFCD16883063
CAS Number: 1027345-07-8
UN: 3265
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR