2-Amino-6-methoxypyridine 98%

Supplier: Ambeed
Warning

A187643-1G A187643-25G A187643-5G A187643-10G A187643-100G
76998-892EA 9 USD
76998-892 76998-894 76998-896 77550-202 77671-660
2-Amino-6-methoxypyridine 98%
2-Amino-6-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₈N₂O
MDL Number: MFCD04972542
CAS Number: 17920-35-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR