2-Hydrazino-4-methylpyridine 95%

Supplier: Ambeed
Warning

Synonyms: (4-Methylpyridin-2-yl)hydrazine, 2-Hydrazino-4-picoline, 2-Hydrazinyl-4-methylpyridine

A187550-250MG A187550-5G
77029-698EA 40.32 USD
77029-698 77671-658
2-Hydrazino-4-methylpyridine 95%
2-Hydrazino-4-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₉N₃
MW: 123.156 g/mol
MDL Number: MFCD09863246
CAS Number: 4931-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR