6-Chloro-2-methylpyridin-3-ol 98%
Supplier: Ambeed
|
Warning
|
A187416-10G
A187416-1G
A187416-250MG
A187416-5G
77012-158EA
625.94
USD
77012-158
77012-160
77012-162
77012-164
6-Chloro-2-methylpyridin-3-ol 98%
6-Chloro-2-methylpyridin-3-ol
Precaución: for Research Use Only
Formula:
C₆H₆ClNO MW: 143.571 g/mol |
MDL Number:
MFCD16610482 CAS Number: 218770-02-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...