(S)-2-(Dimethylamino)propanoic acid 97%

Supplier: Ambeed
Warning

Synonyms: Nα,Nα-DiMe-Ala-OH

A187147-250MG A187147-1G A187147-5G A187147-100MG
76985-976EA 72.15 USD
76985-976 77567-738 77611-816 77683-479
(S)-2-(Dimethylamino)propanoic acid 97%
(S)-2-(Dimethylamino)propanoic acid

Precaución: For research use only.

Formula: C₅H₁₁NO₂
MW: 117.14 g/mol
MDL Number: MFCD00154416
CAS Number: 2812-31-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR