2,6-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile 97%
Supplier: Ambeed
|
Warning
|
A187117-100MG
A187117-10G
A187117-1G
A187117-250MG
A187117-5G
76985-966EA
15
USD
76985-966
76985-968
76985-970
76985-972
76985-974
2,6-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile 97%
2,6-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
Precaución: for Research Use Only
Formula:
C₁₃H₁₄BF₂NO₂ |
MDL Number:
MFCD22419275 CAS Number: 1003298-73-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...