Methyl-3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate, Methyl 3,4-dihydro-3-oxo-2H-1,4-benzothiazine-6-carboxylate

A187114-1G A187114-250MG A187114-25G A187114-5G
76998-882EA 13.8 USD
76998-882 76998-884 76998-886 76998-888
Methyl-3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate 95%
Methyl-3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₉NO₃S
MW: 223.25 g/mol
MDL Number: MFCD00449104
CAS Number: 188614-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR