2,3-Difluoro-4-(trifluoromethyl)benzonitrile 98%

Supplier: Ambeed
Danger

A187025-100MG A187025-1G A187025-250MG
76985-956EA 61 USD
76985-956 76985-958 76985-960
2,3-Difluoro-4-(trifluoromethyl)benzonitrile 98%
2,3-Difluoro-4-(trifluoromethyl)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₂F₅N
MDL Number: MFCD00236258
CAS Number: 240122-24-1
UN: 3276
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR