5-Chloro-2-methylpyrimidin-4-amine 95%

Supplier: Ambeed
Warning

A186961-100MG A186961-1G
77012-152EA 41.38 USD
77012-152 77012-154
5-Chloro-2-methylpyrimidin-4-amine 95%
5-Chloro-2-methylpyrimidin-4-amine

Precaución: for Research Use Only

Formula: C₅H₆ClN₃
MW: 143.57 g/mol
MDL Number: MFCD02728621
CAS Number: 438249-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR