5-Methoxy-2,2-dimethylchroman-4-one 95%

Supplier: Ambeed
Warning

A186918-100MG A186918-1G A186918-250MG
77012-146EA 46.68 USD
77012-146 77012-148 77012-150
5-Methoxy-2,2-dimethylchroman-4-one 95%
5-Methoxy-2,2-dimethylchroman-4-one

Precaución: for Research Use Only

Formula: C₁₂H₁₄O₃
MW: 206.24 g/mol
MDL Number: MFCD15527104
CAS Number: 98910-61-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR