(S)-2-Methoxypropan-1-amine hydrochloride 98%
Supplier: Ambeed
|
Warning
|
A186853-100MG
A186853-1G
A186853-250MG
76985-948EA
49
USD
76985-948
76985-950
76985-952
(S)-2-Methoxypropan-1-amine hydrochloride 98%
(S)-2-Methoxypropan-1-amine hydrochloride
Precaución: for Research Use Only
Formula:
C₄H₁₂ClNO |
MDL Number:
MFCD18381951 CAS Number: 907544-43-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...