3,5-Dichloropyridine 98%

Supplier: Ambeed
Danger

A186736-5G A186736-100G A186736-25G
76998-872EA 8.49 USD
76998-872 76998-874 77600-514 77600-516
3,5-Dichloropyridine 98%
3,5-Dichloropyridine

Precaución: for Research Use Only

Formula: C₅H₃Cl₂N
MDL Number: MFCD00006376
CAS Number: 2457-47-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR