(6-Ethoxy-4-methylpyridin-3-yl)boronic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2-Ethoxy-4-methylpyridine-5-boronic acid

A186672-1G A186672-250MG A186672-5G
76985-924EA 199.45 USD
76985-924 76985-926 76985-928
(6-Ethoxy-4-methylpyridin-3-yl)boronic acid 95%
(6-Ethoxy-4-methylpyridin-3-yl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₁₂BNO₃
MW: 181.00 g/mol
MDL Number: MFCD07374995
CAS Number: 1256355-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR