3,5-Dichloropyrazine-2-carboxamide 98%

Supplier: Ambeed
Warning

A186465-100MG A186465-1G A186465-250MG A186465-25G A186465-5G
76985-912EA 7.43 USD
76985-912 76985-914 76985-916 76985-918 77611-804
3,5-Dichloropyrazine-2-carboxamide 98%
3,5-Dichloropyrazine-2-carboxamide

Precaución: for Research Use Only

Formula: C₅H₃Cl₂N₃O
MW: 192.003 g/mol
MDL Number: MFCD10000837
CAS Number: 312736-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR