Methyl-2-hydroxy-2,2-di(thiophen-2-yl)acetate 96%

Supplier: Ambeed
Warning

A186386-100G A186386-10G A186386-1G A186386-25G A186386-5G
76998-852EA 148 USD
76998-852 76998-854 76998-856 76998-858 77671-634
Methyl-2-hydroxy-2,2-di(thiophen-2-yl)acetate 96%
Methyl-2-hydroxy-2,2-di(thiophen-2-yl)acetate
Formula: C₁₁H₁₀O₃S₂
MDL Number: MFCD08458380
CAS Number: 26447-85-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR