Ethyl 2-(2-iodophenyl)acetate 97%

Supplier: Ambeed
Warning

A186282-1G A186282-250MG A186282-5G
77012-130EA 18 USD
77012-130 77012-132 77012-134
Ethyl 2-(2-iodophenyl)acetate 97%
Ethyl 2-(2-iodophenyl)acetate

Precaución: for Research Use Only

Formula: C₁₀H₁₁IO₂
MDL Number: MFCD00099258
CAS Number: 90794-29-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR