Methyl 3,5-dihydroxy-4-iodobenzoate 95%
Supplier: Ambeed
|
Warning
|
A186050-100MG
A186050-1G
A186050-250MG
77012-124EA
5
USD
77012-124
77012-126
77012-128
Methyl 3,5-dihydroxy-4-iodobenzoate 95%
Methyl 3,5-dihydroxy-4-iodobenzoate
Precaución: for Research Use Only
Formula:
C₈H₇IO₄ |
MDL Number:
MFCD15071424 CAS Number: 338454-02-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...