4-Iodo-1H-pyrazole-5-carboxylic acid 95%

Supplier: Ambeed
Warning

A186047-100MG A186047-1G A186047-5G A186047-10G
77029-626EA 10 USD
77029-626 77029-628 77029-630 77671-626
4-Iodo-1H-pyrazole-5-carboxylic acid 95%
4-Iodo-1H-pyrazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₄H₃IN₂O₂
MDL Number: MFCD00465280
CAS Number: 6647-93-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR