1,4-Bis(methylamino)-9,10-anthraquinone 97%

Supplier: Ambeed
Warning

A186014-100G A186014-25G A186014-5G
77029-624EA 5.31 USD
77029-624 77600-498 77600-500 77600-502
1,4-Bis(methylamino)-9,10-anthraquinone 97%
1,4-Bis(methylamino)-9,10-anthraquinone

Precaución: for Research Use Only

Formula: C₁₆H₁₄N₂O₂
MDL Number: MFCD00001198
CAS Number: 2475-44-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR