1-(Boc-amino)cyclobutanecarboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: N-BOC-1-Aminocyclobutanecarboxylic acid, Boc-cyclovaline, N-tert-Butoxycarbonyl-1-aminocyclobutanecarboxylic acid, 1-((tert-Butoxycarbonyl)amino)cyclobutanecarboxylic acid, N-Boc-1-aminocyclobutanecarboxylic acid

A185953-100G A185953-10G A185953-1G A185953-25G A185953-5G
76998-832EA 156 USD
76998-832 76998-834 76998-836 76998-838 76998-840
1-(Boc-amino)cyclobutanecarboxylic acid 97%
1-(Boc-amino)cyclobutanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₇NO₄
MDL Number: MFCD02682623
CAS Number: 120728-10-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR