1-(2-Hydroxyethyl)piperidine-4-carbonitrile 95%

Supplier: Ambeed
Warning

A185930-10G A185930-1G A185930-250MG A185930-25G A185930-5G
77012-108EA 446 USD
77012-108 77012-110 77012-112 77012-114 77012-116
1-(2-Hydroxyethyl)piperidine-4-carbonitrile 95%
1-(2-Hydroxyethyl)piperidine-4-carbonitrile

Precaución: for Research Use Only

Formula: C₈H₁₄N₂O
MDL Number: MFCD18373979
CAS Number: 21168-73-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR