2-Bromo-5-methoxyisonicotinic acid 95%

Supplier: Ambeed
Warning

A185908-1G A185908-5G A185908-100MG
76985-902EA 211 USD
76985-902 76985-904 77885-410
2-Bromo-5-methoxyisonicotinic acid 95%
2-Bromo-5-methoxyisonicotinic acid

Precaución: for Research Use Only

Formula: C₇H₆BrNO₃
MDL Number: MFCD18257535
CAS Number: 1256789-55-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR