trans-1-(tert-Butoxycarbonyl)-4-(3-(trifluoromethyl)phenyl)pyrrolidine-3-carboxylic acid 98%

Supplier: Ambeed
Warning

A185885-100MG A185885-250MG
77029-616EA 77.45 USD
77029-616 77029-618
trans-1-(tert-Butoxycarbonyl)-4-(3-(trifluoromethyl)phenyl)pyrrolidine-3-carboxylic acid 98%
trans-1-(tert-Butoxycarbonyl)-4-(3-(trifluoromethyl)phenyl)pyrrolidine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₇H₂₀F₃NO₄
MW: 359.34 g/mol
MDL Number: MFCD06659291
CAS Number: 169248-97-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR