[5,5'-Biisobenzofuran]-1,1',3,3'-tetraone 97%

Supplier: Ambeed
Warning

Synonyms: 5-(1,3-Dioxo-2-benzofuran-5-yl)-2-benzofuran-1,3-dione, 3,3',4,4'-Biphenyltetracarbocylic dianhydride. 4,4'-Biphthalic anhydride

A185806-100G A185806-10G A185806-500G A185806-5G A185806-25G
77029-608EA 32 USD
77029-608 77029-610 77029-612 77029-614 77301-998
[5,5'-Biisobenzofuran]-1,1',3,3'-tetraone 97%
[5,5'-Biisobenzofuran]-1,1',3,3'-tetraone

Precaución: for Research Use Only

Formula: C₁₆H₆O₆
MDL Number: MFCD00039140
CAS Number: 2420-87-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR