Methyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylate hydrochloride 97%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: Methyl 1,2,3,4-tetrahydroisoquinoline-3-carboxylate hydrochloride

A185738-250MG A185738-1G
76985-900EA 20 USD
76985-900 77683-473
Methyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylate hydrochloride 97%, Ambeed.Inc
Methyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylate hydrochloride

Precaución: For research use only.

Formula: C₁₁H₁₄ClNO₂
MW: 227.69 g/mol
MDL Number: MFCD00157120
CAS Number: 146074-43-3
New Product

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR