Diphenylmethylene-glycine benzyl ester 95%

Supplier: Ambeed
Warning

Synonyms: N-(Diphenylmethylene) glycine benzyl ester

A185707-1G A185707-25G A185707-5G A185707-100G A185707-10G
77029-600EA 6.37 USD
77029-600 77029-602 77029-604 77600-496 77667-546
Diphenylmethylene-glycine benzyl ester 95%
Diphenylmethylene-glycine benzyl ester

Precaución: for Research Use Only

Formula: C₂₂H₁₉NO₂
MW: 329.3900 g/mol
MDL Number: MFCD08062206
CAS Number: 81477-91-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR