Methyl 2,6-difluoro-4-methoxybenzoate 97%

Supplier: Ambeed
Warning

A185665-100MG A185665-10G A185665-1G A185665-250MG A185665-5G
76985-890EA 25.47 USD
76985-890 76985-892 76985-894 76985-896 76985-898
Methyl 2,6-difluoro-4-methoxybenzoate 97%
Methyl 2,6-difluoro-4-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₈F₂O₃
MW: 202.155 g/mol
MDL Number: MFCD10566175
CAS Number: 84937-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR