3-Oxo-3,4-dihydro-2H-1,4-benzoxazine-7-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 3-Oxo-3,4-dihydro-2H-benzo[b][1,4]oxazine-7-carboxylic acid

A185493-100MG A185493-1G A185493-5G
76998-824EA 43.5 USD
76998-824 76998-826 77550-186
3-Oxo-3,4-dihydro-2H-1,4-benzoxazine-7-carboxylic acid 95%
3-Oxo-3,4-dihydro-2H-1,4-benzoxazine-7-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₇NO₄
MW: 193.16 g/mol
MDL Number: MFCD07774185
CAS Number: 214848-62-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR