(S)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)-4-(tert-butoxycarbonyl)piperazine-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A185230-100MG A185230-1G A185230-250MG
76985-868EA 45.62 USD
76985-868 76985-870 76985-872
(S)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)-4-(tert-butoxycarbonyl)piperazine-2-carboxylic acid 95%
(S)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)-4-(tert-butoxycarbonyl)piperazine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₂₅H₂₈N₂O₆
MW: 452.50 g/mol
MDL Number: MFCD01860733
CAS Number: 244132-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR