2,4-Dichloro-6-methyl-5-nitropyrimidine 98%

Supplier: Ambeed
Warning

A185223-100G A185223-10G A185223-1G A185223-25G A185223-5G
76985-858EA 201.58 USD
76985-858 76985-860 76985-862 76985-864 76985-866
2,4-Dichloro-6-methyl-5-nitropyrimidine 98%
2,4-Dichloro-6-methyl-5-nitropyrimidine

Precaución: for Research Use Only

Formula: C₅H₃Cl₂N₃O₂
MW: 208.0000 g/mol
MDL Number: MFCD00023196
CAS Number: 13162-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR