2-Iodo-3-methoxyaniline 95%

Supplier: Ambeed
Warning

A185153-250MG A185153-1G A185153-100MG A185153-5G
76985-856EA 28 USD
76985-856 77611-794 77890-159 77890-160
2-Iodo-3-methoxyaniline 95%
2-Iodo-3-methoxyaniline

Precaución: for Research Use Only

Formula: C₇H₈INO
MDL Number: MFCD11848601
CAS Number: 98991-09-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR