2H-Pyran-3(6H)-one 97%

Supplier: Ambeed
Warning

A185070-100MG A185070-250MG
76985-852EA 97 USD
76985-852 76985-854
2H-Pyran-3(6H)-one 97%
2H-Pyran-3(6H)-one

Precaución: for Research Use Only

Formula: C₅H₆O₂
MDL Number: MFCD07186238
CAS Number: 98166-23-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR