2-Methylindan-1-one 95%
Supplier: Ambeed
|
Warning
|
A184965-1G
A184965-250MG
A184965-25G
A184965-5G
77012-082EA
18
USD
77012-082
77012-084
77012-086
77012-088
2-Methylindan-1-one 95%
2-Methylindan-1-one
Precaución: for Research Use Only
Formula:
C₁₀H₁₀O |
MDL Number:
MFCD00192303 CAS Number: 17496-14-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...