3-Amino-1,6-dibromoisoquinoline 95%

Supplier: Ambeed
Warning

Synonyms: 1,6-Dibromoisoquinolin-3-amine

A184943-100MG A184943-1G
77029-596EA 71 USD
77029-596 77029-598
3-Amino-1,6-dibromoisoquinoline 95%
3-Amino-1,6-dibromoisoquinoline

Precaución: for Research Use Only

Formula: C₉H₆Br₂N₂
MW: 301.9653 g/mol
MDL Number: MFCD09999214
CAS Number: 925672-85-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR