Methyl 5-chloro-1H-indole-6-carboxylate 98%

Supplier: Ambeed
Warning

A184633-100MG A184633-250MG
77029-588EA 106.09 USD
77029-588 77029-590
Methyl 5-chloro-1H-indole-6-carboxylate 98%
Methyl 5-chloro-1H-indole-6-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₈ClNO₂
MW: 209.62 g/mol
MDL Number: MFCD11976070
CAS Number: 1245643-61-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR