Methyl 2,4-dihydroxypyrimidine-5-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate

A184596-100G A184596-10G A184596-1G A184596-25G A184596-5G
76985-842EA 238.71 USD
76985-842 76985-844 76985-846 76985-848 76985-850
Methyl 2,4-dihydroxypyrimidine-5-carboxylate 95%
Methyl 2,4-dihydroxypyrimidine-5-carboxylate

Precaución: for Research Use Only

Formula: C₆H₆N₂O₄
MW: 170.123 g/mol
MDL Number: MFCD25542372
CAS Number: 42821-92-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR