1-Benzhydrylazetidin-3-ol 98%

Supplier: Ambeed
Warning

Synonyms: 1-(Diphenylmethyl)azetidin-3-ol

A184310-1000G A184310-100G A184310-10G A184310-1G A184310-25G A184310-500G A184310-5G
76998-786EA 221.73 USD
76998-786 76998-788 76998-790 76998-792 76998-794 76998-796 76998-798
1-Benzhydrylazetidin-3-ol 98%
1-Benzhydrylazetidin-3-ol

Precaución: for Research Use Only

Formula: C₁₆H₁₇NO
MW: 239.32 g/mol
MDL Number: MFCD00205109
CAS Number: 18621-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR