1-Boc-2-methyl-D-proline 97%

Supplier: Ambeed
Warning

Synonyms: (R)-1-(tert-Butoxycarbonyl)-2-methylpyrrolidine-2-carboxylic acid, Boc-D-α-Me-Pro-OH

A184247-10G A184247-1G A184247-250MG A184247-5G A184247-100G A184247-25G
76985-832EA 31 USD
76985-832 76985-834 76985-836 76985-838 77885-407 77885-408
1-Boc-2-methyl-D-proline 97%
1-Boc-2-methyl-D-proline

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₄
MW: 229.28 g/mol
MDL Number: MFCD06797554
CAS Number: 166170-15-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR