(R)-Chroman-2-ylmethanamine hydrochloride 95%

Supplier: Ambeed
Warning

A184148-1G A184148-250MG A184148-5G
76985-826EA 45 USD
76985-826 76985-828 76985-830
(R)-Chroman-2-ylmethanamine hydrochloride 95%
(R)-Chroman-2-ylmethanamine hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₄ClNO
MW: 199.68 g/mol
MDL Number: MFCD01863317
CAS Number: 437763-66-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR